Learn More
Waters Corp Tubing Kit, Stainless Steel, Replacement, 515 Isocratic Pump
Catalog No. 502391191 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides a replacement set of stainless steel tubing assemblies for the 515 Isocratic Pump, including all the external tubing assemblies that would need to be replaced.
  • Replacement set of stainless steel tubing assemblies
  • Designed for 515 Isocratic Pump
  • Contains all external tubing assemblies
  • Suitable for HPLC systems
  • Can be replaced as required
Catalog No. 50-239-1191 Supplier Waters Corp Supplier No. 700002680
This kit provides a replacement set of stainless steel tubing assemblies for the 515 Isocratic Pump, including all the external tubing assemblies that would need to be replaced.
  • Replacement set of stainless steel tubing assemblies
  • Designed for 515 Isocratic Pump
  • Contains all external tubing assemblies
  • Suitable for HPLC systems
  • Can be replaced as required