Learn More
Waters Corp Makeup Pump Installation Kit, 515 Pump, UPC² System
Catalog No. 502391634 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit facilitates the use of a 515 pump as a makeup pump within an ACQUITY UPC2 System. It contains all essential components for installation, such as an outlet tube assembly, restrictor, check valve assembly, drip tray, and various other tubing and fittings.
  • Union, SST, 0.020 ID, 10-32 thread
  • Restrictor tube
  • Check valve and bracket assembly
  • Outlet tube
  • Drip tray
  • Elbow, 1/8 NPT to barb, modified
  • Tubing, TYGON, 5/16 ID × 7/16 OD, 72 in.
  • Foot with washer, 0.75 in. tall, 0.60 in. OD
  • Screw, socket head, M4 × 40
  • Tape, Teflon sealant, 6 in.
  • Key, Torx T20
Catalog No. 50-239-1634 Supplier Waters Corp Supplier No. 205001021
This kit facilitates the use of a 515 pump as a makeup pump within an ACQUITY UPC2 System. It contains all essential components for installation, such as an outlet tube assembly, restrictor, check valve assembly, drip tray, and various other tubing and fittings.
  • Union, SST, 0.020 ID, 10-32 thread
  • Restrictor tube
  • Check valve and bracket assembly
  • Outlet tube
  • Drip tray
  • Elbow, 1/8 NPT to barb, modified
  • Tubing, TYGON, 5/16 ID × 7/16 OD, 72 in.
  • Foot with washer, 0.75 in. tall, 0.60 in. OD
  • Screw, socket head, M4 × 40
  • Tape, Teflon sealant, 6 in.
  • Key, Torx T20