Learn More
Sino Biological Recombinant Mouse VISTA/B7-H5 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 51550M08H50
Encompass_Preferred
A DNA sequence encoding the mouse C10orf54 (NP_083008.1) (Met1-Ala191) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3113