Learn More
Sino Biological Recombinant Mouse PLA2G7/PAF-AH Protein (His Tag)
Supplier:  Sino Biological 51562M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Pla2g7 (NP_038765.2) (Met1-Asn440) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3117