Learn More
Sino Biological Recombinant Mouse PRKAR1A Protein (His Tag)
Supplier:  Sino Biological 51743M08B100
Encompass_Preferred
A DNA sequence encoding the mouse PRKAR1A (NP_068680.1) (Met1-Val381) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3120