Learn More
Sino Biological Recombinant Mouse Peroxiredoxin 2 Protein (His Tag)
Supplier:  Sino Biological 51862M07B100
Encompass_Preferred
A DNA sequence encoding the mouse PRDX2 (NP_035693.3) (Met1-Asn198) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3122