Learn More
Bellco Glass, Inc. 51x51cm Flask Tray,250mL
SDP
Catalog No. 501946160
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 16 Dimensions: 51x51cm Size: 250ml. MADE IN THE USA
Catalog No. 50-194-6160 Supplier Bellco Glass, Inc. Supplier No. 774425016
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps. Flask positions: 16 Dimensions: 51x51cm Size: 250ml. MADE IN THE USA