Learn More
AnaSpec SensoLyte 520 ADAM10 Activity Assay Kit
Catalog No. NC1371985
Encompass
Sequence: ELAGIGILTV. Molecular Weight 985.8. ADAM10 has been shown to be an effective α-secretase in vitro and in vivo. This enzyme acts as a sheddase to cleave cell surface proteins involved in neuropathology, inflammatory response and tumor progression.
Catalog No. NC1371985 Supplier AnaSpec Supplier No. AS72226
Sequence: ELAGIGILTV. Molecular Weight 985.8. ADAM10 has been shown to be an effective α-secretase in vitro and in vivo. This enzyme acts as a sheddase to cleave cell surface proteins involved in neuropathology, inflammatory response and tumor progression.