Learn More
AnaSpec 520 MMP FRET Substrate I, Sensitive FRET Assay Tool, High Purity ≥95%, 0.1 mg
Catalog No. NC3672593
Encompass
The 520 MMP FRET Substrate I is a sensitive fluorogenic peptide specially designed for detecting Matrix Metalloproteinases (MMPs). Its unique FRET mechanism allows rapid enzymatic activity measurements, particularly for MMP-13. This substrate excels in research areas like cancer and apoptosis, offering a reliable way to quantify protease activity with clear fluorescent readouts, differentiate enzymatic activity, and ensure reliable results for scientific studies.
Catalog No. NC3672593 Supplier AnaSpec Supplier No. AS6056801
The 520 MMP FRET Substrate I is a sensitive fluorogenic peptide specially designed for detecting Matrix Metalloproteinases (MMPs). Its unique FRET mechanism allows rapid enzymatic activity measurements, particularly for MMP-13. This substrate excels in research areas like cancer and apoptosis, offering a reliable way to quantify protease activity with clear fluorescent readouts, differentiate enzymatic activity, and ensure reliable results for scientific studies.