Learn More
Sino Biological Recombinant Mouse IVD Protein (His Tag)
Supplier:  Sino Biological 52067M07B100
Encompass_Preferred
A DNA sequence encoding the mouse IVD (NP_062800.1) (His31-Arg424) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3132