Learn More
Sino Biological Recombinant Mouse GFOD2 Protein (His Tag)
Supplier:  Sino Biological 52216M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Gfod2 (NP_081745.1) (Met1-Leu385) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3138