Learn More
Sino Biological Recombinant Mouse SMPDL3A Protein (His Tag)
Supplier:  Sino Biological 52271M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Smpdl3a (NP_065586.3) (Met1-Leu445) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3140