Learn More
Sino Biological Recombinant Mouse PIK3IP1 Protein (His Tag)
Supplier:  Sino Biological 52429M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Pik3ip1 (NP_835362.2) (Met1-Leu170) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3144