Learn More
eMolecules​ 52488-36-5 | 4-Bromoindole | Ambeed | MFCD00671502 | 196.047 | C8H6BrN | 97.000 | Brc1cccc2[nH]ccc12 | 10g | 552544834
Catalog No. 5000567191 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Bromoindole | Ambeed | 52488-36-5 | MFCD00671502 | 196.047 | C8H6BrN | 97.000 | Brc1cccc2[nH]ccc12 | 10g | 552544834
Catalog No. 50-005-67191 Supplier eMolecules​ Supplier No. 552544834
4-Bromoindole | Ambeed | 52488-36-5 | MFCD00671502 | 196.047 | C8H6BrN | 97.000 | Brc1cccc2[nH]ccc12 | 10g | 552544834