Learn More
Assay Technology 525 TraceAir® II Organic Vapor MonitorThe 525 Organic Vapor Monitor is our fastest sampling rate badge, which means it is our most sensitive badge with the lowest reporting limits. Use this badge for monitoring chemicals with a low PEL
SDP
Catalog No. NC2964939
Encompass_Preferred
525 TraceAir® II Organic Vapor MonitorThe 525 Organic Vapor Monitor is our fastest sampling rate badge, which means it is our most sensitive badge with the lowest reporting limits. Use this badge for monitoring chemicals with a low PEL (10 ppm or less), STEL monitoring, or for indoor air quality situations.The 525 TraceAir II badge replaced our 521 badge. The design was updated from TraceAir to TraceAir II
Catalog No. NC2964939 Supplier Assay Technology Supplier No. X5251AT
525 TraceAir® II Organic Vapor MonitorThe 525 Organic Vapor Monitor is our fastest sampling rate badge, which means it is our most sensitive badge with the lowest reporting limits. Use this badge for monitoring chemicals with a low PEL (10 ppm or less), STEL monitoring, or for indoor air quality situations.The 525 TraceAir II badge replaced our 521 badge. The design was updated from TraceAir to TraceAir II