Learn More
CELLTREAT Scientific Products 525cm2 Tissue Culture Flask, 3-Layer - Vent Cap, Sterile
SDP
Catalog No. 5000144358
Encompass_Preferred
Optimize time, incubator space, and resources with tissue culture-treated Multiple Layer FlasksCompact design shares the same footprint as 182cm2 flasksFlasks are available with 3 or 5 layers, and may be stacked for even more space saving
Catalog No. 50-001-44358 Supplier CELLTREAT Scientific Products Supplier No. 229323
Optimize time, incubator space, and resources with tissue culture-treated Multiple Layer FlasksCompact design shares the same footprint as 182cm2 flasksFlasks are available with 3 or 5 layers, and may be stacked for even more space saving