Learn More
Sino Biological Recombinant Mouse IL1RAP/IL-1RAcP Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 52657M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Il1rap (NP_598864.1) (Met1-Lys350) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3150