Learn More
Sino Biological Recombinant Mouse MMP-2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 52740M07H100
Encompass_Preferred
A DNA sequence encoding the mouse Mmp2 (NP_032636.1) (Met409-Cys662) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3152