Learn More
Savillex Corporation Savillex 53 mm Closure, Leakproof Seal, High-Purity PFA Material, 53 mm
SDP
Catalog No. NC1921371
Encompass
Savillex 53 mm Closure is designed for high-purity sample storage, offering leakproof protection and exceptional chemical resistance. Ideal for trace metal analysis, it features external grips for easy handling and a wide temperature range of -328°F to 500°F (-200°C to 260°C) making it versatile for various laboratory environments. Its unique design ensures sample integrity, setting it apart from competitors.
Catalog No. NC1921371 Supplier Savillex Corporation Supplier No. 60005301
Savillex 53 mm Closure is designed for high-purity sample storage, offering leakproof protection and exceptional chemical resistance. Ideal for trace metal analysis, it features external grips for easy handling and a wide temperature range of -328°F to 500°F (-200°C to 260°C) making it versatile for various laboratory environments. Its unique design ensures sample integrity, setting it apart from competitors.