Learn More
MSC IPG Tape, Nat Rubber, 1.9 mil, 2x110 yds, Tan, 36/Case
Catalog No. 501918500
Encompass
IPG Series 500 natural rubber packaging tape provides a strong, secure bond for demanding packaging and shipping applications. Its aggressive adhesive ensures reliable sealing on high-recycled content, rough, ultra-smooth, or heavily inked cartons, and performs well in dusty, humid, or extreme temperature environments.
  • Features 1.9 mil thickness for medium-weight box sealing
  • Consistent, reliable sealing performance on highly-recycled content cartons
  • Pressure-sensitive natural rubber adhesive with biaxially oriented polypropylene (BOPP) backing
  • Elongation of 140% for flexibility
  • Temperature use range of 32 to 120 degrees F
  • Coreless design reduces waste
  • Constructed from polypropylene (PP) material
  • Medium duty grade for versatile use
Catalog No. 50-191-8500 Supplier MSC Supplier No. 39580907
IPG Series 500 natural rubber packaging tape provides a strong, secure bond for demanding packaging and shipping applications. Its aggressive adhesive ensures reliable sealing on high-recycled content, rough, ultra-smooth, or heavily inked cartons, and performs well in dusty, humid, or extreme temperature environments.
  • Features 1.9 mil thickness for medium-weight box sealing
  • Consistent, reliable sealing performance on highly-recycled content cartons
  • Pressure-sensitive natural rubber adhesive with biaxially oriented polypropylene (BOPP) backing
  • Elongation of 140% for flexibility
  • Temperature use range of 32 to 120 degrees F
  • Coreless design reduces waste
  • Constructed from polypropylene (PP) material
  • Medium duty grade for versatile use