Learn More
MSC Tape Logic Packing Tape, Clear, 36/Pk.
Supplier:  MSC 39589577
Encompass
Natural Rubber Carton Sealing Tape provides the most consistent, reliable box closure and the most aggressive adhesive of any pressure-sensitive tape. It quickly penetrates and grips corrugated surfaces, reducing 'rub down' time.
  • Most consistent reliable box closure
  • Most aggressive adhesive of any pressure-sensitive tape
  • Quickly penetrates and grips corrugated
  • Moisture resistant, tolerates humid and damp conditions
  • Largest application temperature range: 0°F – 150°F
  • PVC film backing provides lower stretch
  • Maximum tear and puncture resistance
Catalog No. 50-191-9344