Learn More
MSC Tape Logic Packing Tape, Clear, Rubber, 1 ea
Catalog No. 501919345
Encompass
Tape Logic natural rubber carton sealing tape provides a consistent and reliable box closure with a highly aggressive adhesive. It quickly penetrates and grips corrugated surfaces, is moisture resistant, and tolerates a wide range of temperatures.
  • Quickly penetrates and grips corrugated surfaces
  • Moisture resistant, tolerating humid and damp conditions
  • Wide application temperature range of 0° to 150° F
  • Polyvinyl chloride film backing for lower stretch and maximum tear and puncture resistance
  • Has 2.1 mil thickness
  • Offers 25 lb/in tensile strength
  • Features 3 inch width and 55 yd length
Catalog No. 50-191-9345 Supplier MSC Supplier No. 39589585
Tape Logic natural rubber carton sealing tape provides a consistent and reliable box closure with a highly aggressive adhesive. It quickly penetrates and grips corrugated surfaces, is moisture resistant, and tolerates a wide range of temperatures.
  • Quickly penetrates and grips corrugated surfaces
  • Moisture resistant, tolerating humid and damp conditions
  • Wide application temperature range of 0° to 150° F
  • Polyvinyl chloride film backing for lower stretch and maximum tear and puncture resistance
  • Has 2.1 mil thickness
  • Offers 25 lb/in tensile strength
  • Features 3 inch width and 55 yd length