Learn More
Sino Biological Recombinant Mouse Calnexin Protein (His Tag)
Supplier:  Sino Biological 53126M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Canx (NP_031623.1) (Met1-Pro482) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3170