Learn More
Sino Biological Recombinant Mouse OX40 Ligand/TNFSF4 Protein (rFc Tag)
Supplier:  Sino Biological 53582M15H50
Encompass_Preferred
A DNA sequence encoding the mouse Tnfsf4 ( NP_033478.1) (Gln49-Leu198) was expressed with the Fc region of rabbit IgG at the N-terminus..
Catalog No. 50-162-3177