Learn More
Sino Biological Recombinant Mouse RGMB Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 53675M08H50
Encompass_Preferred
A DNA sequence encoding the mouse RGMB (NP_848730.2) (Met1-Cys415) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3179