Learn More
Sino Biological Recombinant Mouse CD44 Protein (His Tag)
Supplier:  Sino Biological 53953M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD44 (NP_001034240.1) (Met1-Thr224) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3180