Learn More
eMolecules​ 543-27-1 | Isobutyl chloroformate | Oakwood Chemicals | MFCD00000642 | 136.580 | C5H9ClO2 | 99.000 | CC(C)COC(Cl)=O | 100g | 480133329
Catalog No. NC3924666 Shop All eMolecules Inc. Products
Encompass_Preferred
Isobutyl chloroformate | Oakwood Chemicals | 543-27-1 | MFCD00000642 | 136.580 | C5H9ClO2 | 99.000 | CC(C)COC(Cl)=O | 100g | 480133329
Catalog No. NC3924666 Supplier eMolecules​ Supplier No. 480133329
Isobutyl chloroformate | Oakwood Chemicals | 543-27-1 | MFCD00000642 | 136.580 | C5H9ClO2 | 99.000 | CC(C)COC(Cl)=O | 100g | 480133329