Learn More
INTERWORLD HIGHWAY LLC Mitutoyo 543-395B - ABSOLUTE Digimatic Indicator ID-C, Inch/Metric, Stem 8mm, Flat Back
SDP
Catalog No. NC3667705
Encompass
Order No. (Model): 543-395B (ID-C112CMXB)Max Range: .5in / 12.7mmStem Diameter: 8mmThread Type: M2.5 x 0.45Back Type: Flat BackResolution: (Switchable) .00005in / .001mm (.0001in and .0005in / .01mm)Accuracy: +/- .0001inMeasuring Force: (Low measuring force) 0.4N -0.7NDust/Water Protection Level: IP42Standards/Specifications: ISO/JIS
Catalog No. NC3667705 Supplier INTERWORLD HIGHWAY LLC Supplier No. IDC112CMXB
Order No. (Model): 543-395B (ID-C112CMXB)Max Range: .5in / 12.7mmStem Diameter: 8mmThread Type: M2.5 x 0.45Back Type: Flat BackResolution: (Switchable) .00005in / .001mm (.0001in and .0005in / .01mm)Accuracy: +/- .0001inMeasuring Force: (Low measuring force) 0.4N -0.7NDust/Water Protection Level: IP42Standards/Specifications: ISO/JIS