Learn More
USA Scientific Inc 5430 cent rotary knob
Catalog No. 502745074
Encompass
Eppendorf 5430 microcentrifuge with 30-place rotor, 17,500 rpm, 30,130 x g, 120 V, 60 Hz
Catalog No. 50-274-5074 Supplier USA Scientific Inc Supplier No. 40620511
Add to Cart
Add to Cart
Request a Quote
Eppendorf 5430 microcentrifuge with 30-place rotor, 17,500 rpm, 30,130 x g, 120 V, 60 Hz