Learn More
Hach Company Hach Duraprene pump tubing, 0.045 in (1.143 mm) ID, Red/Red, pkg. of 6
Catalog No. NC9442677 Shop All Hach Company Products
Encompass
Duraprene pump tubing for pump applications requiring flexible, durable tubing with a 0.045 in (1.143 mm) internal diameter. Supplied as a package of six color-coded tubes to simplify identification and replacement in fluid handling systems.
  • Flexible and durable construction suitable for repetitive pump cycles.
  • 0.045 in (1.143 mm) internal diameter for precise fluid transfer.
  • Color-coded red/red for easy identification.
  • Packaged as a set of six tubes for convenient replacement and maintenance.
  • Designed for pump fluid handling applications.
Catalog No. NC9442677 Supplier Hach Company Supplier No. 54410
Duraprene pump tubing for pump applications requiring flexible, durable tubing with a 0.045 in (1.143 mm) internal diameter. Supplied as a package of six color-coded tubes to simplify identification and replacement in fluid handling systems.
  • Flexible and durable construction suitable for repetitive pump cycles.
  • 0.045 in (1.143 mm) internal diameter for precise fluid transfer.
  • Color-coded red/red for easy identification.
  • Packaged as a set of six tubes for convenient replacement and maintenance.
  • Designed for pump fluid handling applications.