Learn More
Chemglass Life Sciences 55/50 Inner Joint, Un-Cut Drip, on 51mm OD x 2mm W tubing
SDP
Catalog No. CG11723
Encompass_Preferred
55/50 Inner Joint, Un-Cut Drip, on 51mm OD x 2mm W tubing. Drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. CG11723 Supplier Chemglass Life Sciences Supplier No. CG11723
55/50 Inner Joint, Un-Cut Drip, on 51mm OD x 2mm W tubing. Drip tip. Joints are available uncut or with the drip tip being cut at a 45º angle. Tooled on the same tubing sizes as the CG-100 inner joints.