Learn More
Ace Glass, Inc. Soxhlet extractor body, 200mL, 55/50 top, 24/40 bottom, 113mmsiphon height, 50mm ID, use 40x123mm thimble (Ace 6811-24)
SDP
Catalog No. 50973410
Encompass_Preferred
Catalog No. 50-973-410 Supplier Ace Glass, Inc. Supplier No. 673010