Learn More
Chemglass Life Sciences 55/50 Outer Joint, Full Length, on 60mm OD x 2.4mm W tubing
SDP
Catalog No. CG10112
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 55/50 Outer Joint, Full Length, on 60mm OD x 2.4mm W tubing.
Catalog No. CG10112 Supplier Chemglass Life Sciences Supplier No. CG10112
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 55/50 Outer Joint, Full Length, on 60mm OD x 2.4mm W tubing.