Learn More
Ace Glass, Inc. 55/50 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977612
Encompass
55/50 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-612 Supplier Ace Glass, Inc. Supplier No. 1263626
55/50 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.