Learn More
Uline Industrial Trash Liners - 55-60 Gallon 15 Mil, Clear, 100
Supplier:  Uline S13573
Encompass
Premium linear low density - Provides excellent puncture and tear resistance, Handles heavy items and sharp corners without tearing, Strongest trash bags on the market - Made with premium resins, Star seal bottom, FDA compliant, LLDPE Linear Low Density Polyethylene, Performance that is equal to or better than other liners at a thinner mil thickness, Easier handling Conforms better to rigid containers, Meets and exceeds industry accepted dart drop test
Catalog No. NC2468358