Learn More
National Safety Apparel™ 55 Cal TECGEN FR Arc Flash Bib Overalls
SDP
Catalog No. 17001285 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-285 Small
17-001-286 Medium
17-001-287 Large
17-001-288 2X-Large
17-001-289 3X-Large
17-001-290 4X-Large
17-001-291 5X-Large
17-001-292 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-285 Supplier National Safety Apparel™ Supplier No. C453NNBQT55SM32
Add to Cart
Add to Cart
Provides lightweight, durable, heat and flame-resistant solutions. Made of 34% aramid, 33% lyocell, 31% modacrylic, and 2% anti-static materials, Nomex™ is engineered to absorb moisture and dry quickly
  • Arc Rating: 55 cal/cm²
  • CAT 4
  • Fabric/Material: 7.5 oz. TECGEN CC™ OPF Blend Twill / 7 oz. Quilted DuPont™ Nomex™ Lining
  • Lightweight, breathable fabric
  • Inherently flame resistant
  • FR hook & loop fly closure
  • 30% lighter than traditional arc flash suits
  • Heavy duty adjustable, quick disconnect shoulder straps
  • FR hook and loop calf opening for easy boot entry
  • Made in the USA
Specifications
Closure Type Buckle
Color Navy
Material Rayon, Modacrylic, O-Pan, Para-Aramid
Seam Type Stitched
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 55 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable