Learn More
National Safety Apparel™ 55 Cal Tecgen FR Arc Flash Coat
SDP
Catalog No. 17001164 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-164 5X-Large
17-001-157 Small
17-001-158 Medium
17-001-159 Large
17-001-160 X-Large
17-001-161 2X-Large
17-001-162 3X-Large
17-001-163 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-164 Supplier National Safety Apparel™ Supplier No. C043NNBQT555X32
Lightweight and durable arc flash coat is an ideal heat and flame-resistant solution.
  • Arc Rating: 55 cal/cm²
  • CAT 4
  • Fabric/Material: 7.5 oz. TECGEN CC™ OPF Blend Twill / 7 oz. Quilted DuPont™ Nomex™ Lining
  • FR knit wrist cuffs keep sleeves in place Lightweight, breathable fabric
  • Inherently flame resistant
  • FR hook and loop front closure
  • 30% lighter than traditional arc flash suits Made in the USA
Specifications
Closure Type FR Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Rayon, Modacrylic, O-Pan, Para-Aramid
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 55 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269