Learn More
National Safety Apparel™ 55 Cal Tecgen FR Arc Flash Hood
SDP
Catalog No. 17001713 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-713 Color: Navy
1 options
Catalog No. 17-001-713 Supplier National Safety Apparel™ Supplier No. H653NNBQT55HTHAT
DuPont™ Nomex™ Brand Fiber Provides lightweight, durable, heat and flame-resistant solutions. Made of 34% aramid, 33% lyocell, 31% modacrylic, and 2% anti-static materials, Nomex™ is engineered to absorb moisture and dry quickly
  • Arc Rating: 55 cal/cm²
  • CAT 4
  • Fabric/Material: 7.5 oz. TECGEN CC™ OPF Blend Twill / 7 oz. Quilted DuPont™ Nomex™ Lining
  • Highly transparent shield for better color recognition and improved visibility
  • Removable/replaceable anti-fog and anti-abrasion coated faceshield Inherently flame resistant
  • Lightweight, breathable fabric Made in the USA
Specifications
Size Medium
Material Rayon, Modacrylic, O-Pan, Para-Aramid
Color Navy
Seam Type Stitched
Arc Flash Rating 55 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269