Learn More
Labelmaster Inc 55 GAL. EPOXY LINED DRUM
SDP
Catalog No. NC9547536
Encompass_Preferred
This Steel Drum With Closed Head Bolt Ring Closure is constructed of cold rolled carbon steel. The 16/16/16 gauge (1.5/1.5/1.5 mm) steel drum has a 2 in. steel plug with 3/4; vent in top. UN certified for liquids only.
Catalog No. NC9547536 Supplier Labelmaster Inc Supplier No. KM5516REP
This Steel Drum With Closed Head Bolt Ring Closure is constructed of cold rolled carbon steel. The 16/16/16 gauge (1.5/1.5/1.5 mm) steel drum has a 2 in. steel plug with 3/4; vent in top. UN certified for liquids only.