Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ 55 Gallon Lab Pack - Open Head Drum
GREENER_CHOICE

Catalog No. 19038455 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-038-455 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-038-455 Supplier Eagle™ Supplier No. 1656MB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This straight sided drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors. It is durable and lightweight, weather and chemical resistant, and has a white lid with a plastic lever-lock band closure.

  • Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
  • Lab Packs/Open Head Drums - for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
  • Color: Blue
  • Closure Type: Metal Lever Lock
  • Capacity: 55 gallon
  • DOT Rating: 1H2/X200/S 1H2/Y250/S

Specifications

Certifications/Compliance UN/DOT: 1H2/X200/S 1H2Y250/S
Closure Type Metal Lever Lock Lid
Capacity (English) 55 gal.
Capacity (Metric) 217 L
Color Blue
For Use With (Application) Small Non-Damaged Bottles, Cans, and Carboys. 5 Gallon Pails of Non-Leaking Hazardous Materials. NO FREE LIQUIDS
Material HDPE
Finish Poly
Freight Class 300
Head Style Lab Pack
Includes Metal Lever Lock Lid
Warranty 1 Year
Height (English) 36.38 in.
Height (Metric) 92.4 cm
Diameter (English) Inner 22.125 in.
Diameter (English) Opening 20.06 in.
Diameter (English) Outer 22.5 in.
Diameter (Metric) Inner 56.2 cm
Diameter (Metric) Opening 51.2 cm
Diameter (Metric) Outer 57.2 cm
Weight (English) 23.5 lb.
Weight (Metric) 10.7 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.