Learn More
Cary Company 55 Gallon Tight Head Stainless Steel Drum, UN Rated, 2" & 3/4" Fittings (18 gauge)****Minimum order quaatity of 8****
SDP
Catalog No. NC4042020
Encompass
Made from 18 gauge, 304 stainless steel, our UN Rated 55 gallon tight head stainless steel drums (closed head) are an ideal solution for products requiring drum purity and compatibility of materials, depending on their composition. These drums can outperform carbon steel drums in terms of corrosion resistance, tensile strength and reusability. These tight head stainless steel drums feature 2" & 3/4" Tri-Sure stainless steel fittings.****Minimum order quaatity of 8****
Catalog No. NC4042020 Supplier Cary Company Supplier No. 26WST5 MOQ 8
Made from 18 gauge, 304 stainless steel, our UN Rated 55 gallon tight head stainless steel drums (closed head) are an ideal solution for products requiring drum purity and compatibility of materials, depending on their composition. These drums can outperform carbon steel drums in terms of corrosion resistance, tensile strength and reusability. These tight head stainless steel drums feature 2" & 3/4" Tri-Sure stainless steel fittings.****Minimum order quaatity of 8****