Learn More
Uline Steel Drum with Lid - 55 Gallon Open Top Unlined, Black
Catalog No. NC2176859
Encompass
Store and ship liquids or solids,Withstands extreme temperatures of -30 to 250F,Features 2 opening to pour and 34 opening to vent,Open Top Includes removable top and bolt ring
Catalog No. NC2176859 Supplier Uline Supplier No. S10758
Store and ship liquids or solids,Withstands extreme temperatures of -30 to 250F,Features 2 opening to pour and 34 opening to vent,Open Top Includes removable top and bolt ring