Learn More
Accuflow Systems Altafluor 550 PVDF Flex is trade name Kynar Flex -
Supplier:  Accuflow Systems 550050006250
Encompass_Preferred
Altafluor 550 KYNAR FLEX PVDF (Polyvinylidene flouride) is a more flexible version of PVDF. It is suitable for use with flare fittings, is resistant to many commonly used industrial fluids and gases (including ozone), and does not support the growth of fungi. It is preferred for water disinfecting processes such as aquarium, spa, pool and agriculture. Kynar Flex PVDF is approved for use in food contact applications in compliance with FDA regulation CFR 177.2600, NSF: tubing is produced using NSF 51 and NSF certified resin. It is chemically resistant to many indrstrial chemicals and solvents and has low smoke and flame properties and resistant to bacterial and fungal growth.
Catalog No. 50-135-5386