Learn More
Hach Company Hach 5500sc Ammonia Monochloramine Acidic Surfactant Wash, 2 L (for 5500sc analyzer)
Catalog No. NC2381549 Shop All Hach Company Products
Encompass
Cleaning solution formulated for routine maintenance of the 5500sc Ammonia/Monochloramine analyzer. Supplied in a 2 L bottle, this acidic surfactant wash removes fouling and residues from probes and flow cells to help maintain measurement accuracy.
  • Supplied in a 2 L bottle for routine maintenance.
  • Formulated with an acidic surfactant to remove fouling and residues.
  • Designed for online ammonia and monochloramine measurement flow cells and probes.
  • Helps maintain measurement accuracy through regular cleaning.
  • Store in a dry, cool, well-ventilated place; protect from moisture.
  • May be subject to hazardous shipping restrictions.
Catalog No. NC2381549 Supplier Hach Company Supplier No. 25239000
Add to Cart
Add to Cart
Cleaning solution formulated for routine maintenance of the 5500sc Ammonia/Monochloramine analyzer. Supplied in a 2 L bottle, this acidic surfactant wash removes fouling and residues from probes and flow cells to help maintain measurement accuracy.
  • Supplied in a 2 L bottle for routine maintenance.
  • Formulated with an acidic surfactant to remove fouling and residues.
  • Designed for online ammonia and monochloramine measurement flow cells and probes.
  • Helps maintain measurement accuracy through regular cleaning.
  • Store in a dry, cool, well-ventilated place; protect from moisture.
  • May be subject to hazardous shipping restrictions.