Learn More
Hach Company 5500sc Ammonia Monochloramine Maintenance Kit, 2-Channel - for 5500sc Analyzer
Supplier:  Hach Company 9560502
Encompass
The 5500sc Ammonia Monochloramine Maintenance Kit (2 Channel) provides the replacement consumables and routine parts intended for scheduled, annual maintenance of the 5500sc Ammonia Monochloramine Analyzer. It helps maintain analyzer accuracy and reliability by replacing wear items and service components.
  • Intended for annual replacement of consumable components.
  • Designed for two-channel analyzer configurations.
  • Helps maintain analyzer performance and measurement reliability.
  • Includes replacement parts and consumables for routine maintenance.
Catalog No. NC2480347