Learn More
Hach Company Hach 5500sc Ammonia Monochloramine Reagent 2, 1 L
Supplier:  Hach Company 25235000
Encompass
Hach 5500sc Ammonia Monochloramine Reagent 2 is supplied as a 1 L bottle for use with online ammonia and monochloramine analyzers. The reagent is part of the chemistry required to perform reliable ammonia/monochloramine measurements and should be handled and stored according to safety guidelines.
  • Supports measurement of ammonia and monochloramine.
  • Supplied as a 1 L bottle for routine reagent replacement.
  • Requires hazardous shipping and appropriate handling.
  • Store tightly closed in a dry, well-ventilated place.
  • Intended for routine maintenance and continuous monitoring applications.
Catalog No. NC2381544