Learn More
Hach Company Hach 5500sc Ammonia Monochloramine Standard 1 (0 ppm NH3), 2 L
Catalog No. NC2381546 Shop All Hach Company Products
Encompass
The 5500sc Ammonia Monochloramine Standard 1 is a factory-prepared, zero-concentration calibration standard supplied in a 2 L bottle for verification and calibration of ammonia/monochloramine analyzers.
  • provides a 0 ppm NH3 reference for calibration and verification.
  • supplied in a ready-to-use 2 L liquid bottle.
  • reduces analyzer drift and supports measurement accuracy over time.
  • suitable for use with compatible ammonia-monochloramine online analyzers.
  • store tightly closed in a dry, well-ventilated place.
Catalog No. NC2381546 Supplier Hach Company Supplier No. 25237000
The 5500sc Ammonia Monochloramine Standard 1 is a factory-prepared, zero-concentration calibration standard supplied in a 2 L bottle for verification and calibration of ammonia/monochloramine analyzers.
  • provides a 0 ppm NH3 reference for calibration and verification.
  • supplied in a ready-to-use 2 L liquid bottle.
  • reduces analyzer drift and supports measurement accuracy over time.
  • suitable for use with compatible ammonia-monochloramine online analyzers.
  • store tightly closed in a dry, well-ventilated place.