Learn More
Hach Company Hach 5500sc Ammonia Monochloramine Reagent 3, 1 L - for 5500sc AMC analyzer
Catalog No. NC2308731 Shop All Hach Company Products
Encompass
This 1 L reagent (Reagent 3) is formulated for use with the 5500sc Ammonia/Monochloramine online analyzer to support accurate ammonia/monochloramine measurements as part of the analyzer's reagent chemistry sequence.
  • 1 L bottle.
  • Formulated for ammonia/monochloramine parameter.
  • Intended as reagent 3 in an online analyzer chemistry sequence.
  • Supports reliable, routine reagent replacement and maintenance.
Catalog No. NC2308731 Supplier Hach Company Supplier No. 25236000
This 1 L reagent (Reagent 3) is formulated for use with the 5500sc Ammonia/Monochloramine online analyzer to support accurate ammonia/monochloramine measurements as part of the analyzer's reagent chemistry sequence.
  • 1 L bottle.
  • Formulated for ammonia/monochloramine parameter.
  • Intended as reagent 3 in an online analyzer chemistry sequence.
  • Supports reliable, routine reagent replacement and maintenance.