Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Hach 5500sc Silica Analyzer, 2 Channels

Supplier:  Hach Company 5500.KTO.S0.A2U

Encompass

The Hach 5500 sc Silica Analyzer (MPN: 5500.KTO.S0.A2U) is an online colorimetric analyzer for continuous monitoring of dissolved silica (SiO2) in water. It supports two programmable analysis channels, extended reagent life for unattended operation, and predictive diagnostics to reduce downtime.

  • Continuous colorimetric measurement of silica at 815 nm.
  • Measuring range: 0 to 5,000 μg/L as SiO2.
  • Accuracy: 0-500 μg/L ±1% or ±1 μg/L; 500-5,000 μg/L ±5%.
  • Two programmable channels for sequential analysis.
  • Uses 2 L reagent bottles providing up to 90 days of unattended operation (15-minute cycle time).
  • Pressurized reagent delivery and color-coded caps for quick, clean reagent changes.
  • AC power operation and built-in diagnostics to reduce downtime.

Catalog No. NC3091573


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.