Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company KTO: 5500sc Silica Analyser Reagent Set - Hach

Catalog No. NC3443161 Shop All Hach Company Products
Encompass

Complete reagent and standards set designed for use with the 5500sc silica analyser. The set provides reagents, standards, and a citric acid solution for instrument startup and routine operation. The R2 reagent is modified to reduce blue staining in sensitive applications. Items in this set may be subject to hazardous shipping restrictions.

  • Includes silica standard, 2 L
  • Includes silica reagent 3, 1.8 L
  • Includes silica reagent 3 powder, 205 g
  • Includes silica reagent 1, 2 L
  • Includes citric acid reagent solution, 2 L
  • Pack size: one reagent set
  • Intended compatibility: 5500sc silica analyser
  • May require special shipping due to hazardous materials

Catalog No. NC3443161 Supplier Hach Company Supplier No. 25286000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complete reagent and standards set designed for use with the 5500sc silica analyser. The set provides reagents, standards, and a citric acid solution for instrument startup and routine operation. The R2 reagent is modified to reduce blue staining in sensitive applications. Items in this set may be subject to hazardous shipping restrictions.

  • Includes silica standard, 2 L
  • Includes silica reagent 3, 1.8 L
  • Includes silica reagent 3 powder, 205 g
  • Includes silica reagent 1, 2 L
  • Includes citric acid reagent solution, 2 L
  • Pack size: one reagent set
  • Intended compatibility: 5500sc silica analyser
  • May require special shipping due to hazardous materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.